Kimble™ Kontes™ Brand Microflex™ Microscale Kits - Syringe Needles
Nonsterile
Manufacturer:  Kimble™ 8682801801
 View more versions of this product
Catalog No. 11-304-21
Specifications
Description
Needles, Syringe; Kimble-Chase Kontes; For Microflex Microscale Kits; Nonsterile; Gauge: 18; Size: 1 in.; 12/pk
  • Size: 1 in.
  • Sold 12/pk
Description & Specifications
Specifications
0.05 in.
For general purpose dispensing in laboratory applications and for microflex microscale kits
Stainless Steel
18 ga
1.25mm
With an inert plastic luer hub and a blunt end for safety
Yes
Syringe Needle