Please login to your online account to display your discounted pricing

Kimble™ Kontes™ Adsorption/Ion-Exchange Columns


With or without fritted disc

Manufacturer: kimble™  4203000022

 View more versions of this product

Catalog No. K420300-0022

  • / Each
 

Description & Specifications

Specifications

Column Capacity 110mL
For Use With Many separations in adsorption and ion exchange chromatography
Length (Metric) 300mm
Material Borosilicate Glass
Support Material Glass Wool
Diameter (Metric) Inner 22mm
Includes Without fritted disc
Type Adsorption/Ion Exchange Column