Please login to your online account to display your discounted pricing

Kimble™ Kontes™ 47mm Ultra-Ware™ Microfiltration Assembly Component Parts


Components for Ultra-ware™ microfiltration assembly

Manufacturer: kimble™  9537770001

 View more versions of this product

Catalog No. K953777-0001

  • / Each
 

Description & Specifications

Specifications

Material Fritted Glass
For Use With (Equipment) Ultra-ware microfiltration assembly with PTFE-faced fritted glass support
Description Support Base; 47mm