missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
DWK Life Sciences Kimble™ Kontes™ 20mm Flip-Off Button-Top Aluminum Seals, Color Coded
Two-piece, tamper-evident flip-off feature protects target area
Supplier: DWK Life Sciences 73845A20
Description
- Button top is easily removed with the flip of a thumb
- For use with 20mm serum finished bottles and vials
- Designed to secure septa and stoppers to bottles and vials
- Available with blue, red, green and white plastic buttons to help separate and identify samples
Specifications
White | |
Autoclavable | |
Flip-up Button-top Cap |
Aluminum | |
1000 Cs. | |
20 mm |