missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
DWK Life Sciences Kimble™ Kontes™ 20mm Flip-Off Button-Top Aluminum Seals, Color Coded
Two-piece, tamper-evident flip-off feature protects target area
$507.24 - $507.24
Specifications
Autoclavable | Autoclavable |
---|---|
Closure Type | Flip-up Button-top |
Closure Material | Aluminum |
Quantity | 1000 Case |
Thread Size | 20 mm |
Catalog Number | Mfr. No. | Color | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Price | Quantity | |||||
03-340-336
|
DWK Life Sciences
73845A20 |
White |
Case of 1000 Each for $507.24
|
|
03-340-337
|
DWK Life Sciences
73845B20 |
Blue |
Case of 1000 Each for $507.24
|
|
03-340-339
|
DWK Life Sciences
73845D20 |
Green |
Case of 1000 Each for $507.24
|
|
03-340-338
|
DWK Life Sciences
73845C20 |
Red |
Case of 1000 Each for $507.24
|
|
Description
- Button top is easily removed with the flip of a thumb
- For use with 20mm serum finished bottles and vials
- Designed to secure septa and stoppers to bottles and vials
- Available with blue, red, green and white plastic buttons to help separate and identify samples
Specifications
Autoclavable | |
Aluminum | |
20 mm |
Flip-up Button-top | |
1000 Case |