DWK Life Sciences Kimble™ Serological Disposable Pipet
Sterile, color coded
Manufacturer:  DWK Life Sciences K7210210110
 View more versions of this product
Catalog No. 13678253
Description
Description
  • Borosilicate glass
  • Plugged
  • Calibrated
  • Non-Pyrogenic
Specifications
Specifications
0.33 oz.
ASTM E714
Yes
0.1mL
±0.03mL
10mL
Sterile
0.33 oz., 10mL
Orange
Yes
2.5mL
Serological Pipet
Borosilicate Glass
Documents