DWK Life Sciences Kimble™ Serological Disposable Pipet
Sterile, color coded
Manufacturer:  DWK Life Sciences K721025110
 View more versions of this product
Catalog No. 13678252
Description
Description
  • Borosilicate glass
  • Plugged
  • Calibrated
  • Non-Pyrogenic
Specifications
Specifications
0.16 oz.
ASTM E714
Yes
0.1mL
±0.15mL
5mL
Sterile
0.16 oz., 5mL
Blue
Yes
1mL
Serological Pipet
Borosilicate Glass
Documents