missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Kimble™ Screw-Thread N-51A Borosilicate Glass Sample Vials: Without Caps

Low extractable Kimax N-51A glass

$108.00 - $135.50

Specifications

Certifications/Compliance ASTM Type I
Classification B
Color Clear
Material Borosilicate Glass
Type Sample Vial
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Closure Type Volume (English) Capacity (English) Price Quantity  
Catalog Number Mfr. No. Closure Type Volume (English) Capacity (English) Price Quantity  
03-339-19A Kimble™
K609101
Screw cap 1 dr. 1 dram Pack of 144 Each for $108.00
Case of 24 Pack for $2,592.00
03-339-19D Kimble™
K609102
Screw cap 2.16 dr. 2 dram Pack of 144 Each for $135.50
Case of 15 Pack for $2,032.50
Specifications

Description

  • Borosilicate glass sample vials without caps

Compliance

Kimax N-51A glass conforms to ASTM Type 1, Class B requirements.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Description & Specifications

Specifications

ASTM Type I
Clear
Sample Vial
Screw
B
Borosilicate Glass
For general purpose