DWK Life Sciences Kimble™ KIMAX™ Valved Outlet Reservoir Bottles with Tubulation
With detachable, quick-release connector
Supplier:  DWK Life Sciences 14612F2000
 View more versions of this product
Catalog No. 02-973-2A
Requires : Flexible tubing
Description
Description
  • Tooled top, narrow mouth for rubber stopper
  • Graduated with approximate capacity scale
  • Bottom outlet controlled with an 8mm valve and can be used with 0.25 in. (6mm) flexible tubing (not supplied)
Compliance
Meet ASTM™ E 438 Type I, Class A requirements.
Specifications
Specifications
Reservoir Bottle
1.06 in.
0.25 in.
136 mm
2000 mL
Designed to store and discharge liquids via a bottom valve outlet
250 mm
6
0.5 gal.
5.35 in.
27 mm
6 mm
Borosilicate Glass
9.84 in.
Bottom hose outlet
SDS
Documents
Documents
Accessories