missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
DWK Life Sciences Kimble™ KIMAX™ Storage/Media Bottles
With linerless blue polypropylene caps
Supplier: DWK Life Sciences KC143955000
Description
With linerless blue polypropylene caps
- KimCote™ plastic coating helps prevent and contain spills in the event of glassware breakage
- GL 45 screw thread bottles with white graduations and marking spots
- Wide opening for easy access (approx. 30mm I.D.)
- Linerless cap is blue polypropylene and autoclavable (140°C max)
- Replaceable pour ring included
- Ultra-clear for improved sample viewing
- Non-slip surface — wet or dry
Applications:
Pressurized glass, chemical storage, transportation of samples, handling of hazardous chemicals, harmful biologicals and acids
Specifications
Polypropylene | |
330 mm | |
12.9 in. | |
GL 45 | |
5000 mL | |
Ultra Clear | |
1.18 in. | |
Clear drip-free Polypropylene pouring ring | |
Blue | |
Yes | |
Yes |
Media Bottle | |
181 mm | |
Screw Cap | |
169.07 oz., 5000 mL | |
169.07 oz. | |
7.1 in. | |
30 mm | |
Plastic | |
1000 to 4000mL | |
Autoclavable | |
Wide |