missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

DWK Life Sciences Kimble™ KIMAX™ Storage/Media Bottles

With linerless blue polypropylene caps

Manufacturer:  DWK Life Sciences KC14395250

 View more versions of this product

Catalog No. 12-141-321



Description

Description

With linerless blue polypropylene caps

  • KimCote™ plastic coating helps prevent and contain spills in the event of glassware breakage
  • GL 45 screw thread bottles with white graduations and marking spots
  • Wide opening for easy access (approx. 30mm I.D.)
  • Linerless cap is blue polypropylene and autoclavable (140°C max)
  • Replaceable pour ring included
  • Ultra-clear for improved sample viewing
  • Non-slip surface — wet or dry

Applications:

Pressurized glass, chemical storage, transportation of samples, handling of hazardous chemicals, harmful biologicals and acids

Specifications

Specifications

8.45 oz., 250 mL
8.45 oz.
2.7 in.
30mm
Plastic
50 to 200mL
Polypropylene
70 mm
5.4 in.
250 mL
Ultra Clear
1.18 in.
Clear drip-free Polypropylene pouring ring
Blue
GL45
Screw Cap
Yes
138 mm
SDS
Documents
Product Certifications
Promotions

Promotions