missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
DWK Life Sciences Kimble™ KIMAX™ Squibb Funnels with standard taper Stopcock and Stopper
Lower stems made of large-diameter tubing for liquid to drain out completely when stopcock is closed
Supplier: DWK Life Sciences 29048500
Specifications
500 mL | |
ASTM E1096, Type IV | |
Each | |
Glass | |
Squibb separatory funnel |
Borosilicate Glass | |
60mm | |
4; Glass | |
27 |