Please login to your online account to display your discounted pricing
Kimble™ KIMAX™ Soxhlet Apparatus with Allihn Condenser, Extraction tube only
Manufacturer: kimble™  K2407130
 View more versions of this product
Catalog No. 09-553A
  • / Each
Description & Specifications
Specifications
Diameter (English) Inside 1.18 in.
Diameter (Metric) Inside 3cm
Standard Taper Joint Top 34/45
Standard Taper Joint 24/40
For Use With (Equipment) Soxhlet apparatus
Type Extraction Tube