Kimble™ KIMAX™ Soxhlet Apparatus with Allihn Condenser, Condenser only
Manufacturer:  Kimble™ K2402730
 View more versions of this product
Catalog No. 09-551-10A
Specifications
Description
Kimble No.: 24027 30; standard taper Joint: 34/45
  • The soxhlet extractor was originally designed for the extraction of a lipid from a solid material
  • Not limited to the extraction of lipids - typically, a soxhlet extraction is only required where the desired compound has a limited solubility in a solvent and the impurity is insoluble in that solvent
  • If the desired compound has a high solubility in a solvent then a simple filtration can be used to separate the compound from the insoluble substance
Description & Specifications
Specifications
1.18 in.
34/45
Extractor Condenser
3cm
Soxhlet apparatus