DWK Life Sciences Kimble™ KIMAX™ Short Neck Flask with standard taper Joints
Flat Bottom Boiling
Supplier: DWK Life Sciences 25055300
Description
- Short Neck with 24/40
- ASTM E 1403, TI
Specifications
3.4 in. | |
Borosilicate Glass | |
13.5 cm | |
24/40 |
300 mL | |
5.3 in. | |
Short | |
87 mm |