Kimble™ KIMAX™ Organic Chemistry Kits
Include an assortment of items required for basic organic laboratory experiments
Manufacturer:  Kimble™ 3382099
 View more versions of this product
Catalog No. 11-301-20
Specifications Description & Specifications
Specifications
USP Type I and ASTM E438 Type I, Class A
For extraction, reflux, reflux with addition, fractionation and distillation (steam, atmospheric and vacuum), as well as other organic preparations
139.7mm
23 in.
18 in.
Full Scale Organic Chemistry Kit
23 x 18 x 5.5 in. (584.2 x 457.2 x 139.7mm)
5.5 in.
300mm west condenser, 300mm distilling column, a 50mL, 100mL, 200mL, and 300mL single neck round bottom flask, 125mL separatory funnel, 2 hollow glass stoppers, thermometer adapter tube, claisen connecting tube, three-way connecting tube
584.2mm
457.2mm