Please login to your online account to display your discounted pricing
Kimble™ KIMAX™ Organic Chemistry Kits
Include an assortment of items required for basic organic laboratory experiments
Manufacturer: kimble™  2693200000
 View more versions of this product
Catalog No. K269320-0000
  • / Each
Description & Specifications
Specifications
Standard Taper Joint 14/20
Certifications/Compliance USP Type I and ASTM E438 Type I, Class A
Includes Bleed adapter, claisen adapter, condenser, connecting adapter, distillation column, 25mL, 50mL, 100mL, 250mL, 500mL Flask, 125mL PTFE funnel, outlet adapter, stopper, solid glass, vacuum adapter
For Use With (Application) For extraction, reflux, reflux with addition, fractionation, and distillation
Product Type Standard Taper Kit