Please login to your online account to display your discounted pricing
Kimble™ KIMAX™ Nonvacuum Glass Desiccators
Glass desiccators made for durability and efficiency in daily use
$649.53 - $649.53
Specifications
Certifications/Compliance USP Type I, ASTM E438, Type I, Class A
Product Type Nonvacuum Desiccator
Includes Cover
Material Borosilicate Glass
Style Knob top
View More Specs
Includes:  Cover
Requires:  Desiccant plate
Products
Catalog Number Mfr. No. Size Price Quantity    
08-595-6
  Accessories
kimble™
K21050160
Small Each for $649.53
Description & Specifications
Specifications
Certifications/Compliance USP Type I, ASTM E438, Type I, Class A
Product Type Nonvacuum Desiccator
Includes Cover
Material Borosilicate Glass
Style Knob top
  • Jars shaped to hold sample plate (not supplied) above desiccant
  • Bottoms sloped so desiccant can be removed easily
  • Manufactured from 33 expansion borosilicate glass
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.