DWK Life Sciences Kimble™ KIMAX™ Narrow-Mouth Solution Bottles with Color-Coded PTFE standard taper Flathead Stoppers
Plastic-coated for added handling protection; color-coded for easy identification
$534.89 - $1,418.84
Specifications
Certifications/Compliance ASTM E438, Type I, Class A
Closure Material PTFE
Coating Plastic
Closure Type Flat Stopper
View More Specs
Includes: Stoppers
Products 3
Catalog Number Mfr. No. Capacity Closure Color Height (Metric) Height (English) Price Quantity  
Catalog Number Mfr. No. Capacity Closure Color Height (Metric) Height (English) Price Quantity  
12-141-317
View Documents Promotion Details
DWK Life Sciences
KC15097500
16 oz., 500 mL Blue 180 mm 7 in.
Case of 6 Each for $1,189.16
12-141-318
View Documents Promotion Details
DWK Life Sciences
KC150971000
32 oz., 1000 mL Green 215 mm 8.4 in.
Case of 6 Each for $1,418.84
12-141-319
View Documents Promotion Details
DWK Life Sciences
KC150972000
67.62 oz., 2000 mL Green 265 mm 10.4 in.
Case for $534.89
Description
Description
  • KimCote™ plastic coating prevents and contains spills
  • Ultra-clear bottles for improved sample viewing
  • Non-slip surface—wet or dry
  • Autoclavable and chemical resistant
  • With standard taper necks
  • Stoppers color-coded for easy identification
Compliance
Conforms to ASTM™ E438, Type 1, Class A glass
Specifications
Specifications
ASTM E438, Type I, Class A
PTFE
Plastic
Flat Stopper