Kimble™ KIMAX™ Flasks with standard taper Pennyhead Stoppers
Calibrated “To Contain”
Manufacturer:  Kimble™ K2801450
 View more versions of this product
Catalog No. 10-209C
Specifications Description & Specifications
Specifications
Calibrated To Contain
9
14.8cm
50mL
Glass
5.8 in.
±0.05mL
Borosilicate Glass