DWK Life Sciences Kimble™ KIMAX™ Flasks Serialized and Class A Certified with Snap Caps
Calibrated and certified against NIST-Traceable equipment
Supplier:  DWK Life Sciences 28012250
 View more versions of this product
Catalog No. 10-201C
Description
Description
Calibrated and certified against NIST-Traceable equipment
  • Permanent individual serial no.
  • Designed according to ASTM E 288
  • To contain
Specifications
Specifications
Class A, USP Type I, ASTM E438, E288
250 mL
8.8 in.
±0.12 mL
Polyethylene
Borosilicate Glass
22.5 cm
5