Kimble™ KIMAX™ Flasks Serialized and Certified with standard taper Pennyhead Stopper
Manufacturer:  Kimble™ K28017100
 View more versions of this product
Catalog No. 10-212B
Specifications
Description
Flask, Volumetric Class A, Kimax; Serialized/Certified; Permanent individual serial no.; Calibrated and certified against NIST-Traceable equipment; Designed according to ASTM E 288; With Standard Taper stopper No. 13; To Contain; 100mL
  • Permanent individual serial no.
  • Designed according to ASTM E 288
  • To contain
Description & Specifications
Specifications
Glass
7.08 in.
±0.08mL
Borosilicate Glass
13
18cm
100mL