Kimble™ KIMAX™ Flasks Serialized and Certified with standard taper Pennyhead Stopper
Manufacturer:  Kimble™ K280171000
 View more versions of this product
Catalog No. 10-212F
Specifications
Description
Flask, Volumetric Class A, Kimax; Serialized/Certified; Permanent individual serial no.; Calibrated and certified against NIST-Traceable equipment; Designed according to ASTM E 288; With Standard Taper stopper No. 22; To Contain; 1000mL
  • Permanent individual serial no.
  • Designed according to ASTM E 288
  • To contain
Description & Specifications
Specifications
Class A, USP Type I, ASTM E438, E288
22
33.8cm
1000mL
Glass
13.3 in.
±0.3mL
Borosilicate Glass