DWK Life Sciences Kimble™ KIMAX™ Flasks with Graduation Line Etched through Vertical Red Stripe
Calibrated “To Contain” within Class A tolerances
Supplier:  DWK Life Sciences 280131000
 View more versions of this product
Catalog No. 10-248G
Description
Description
  • Meet specifications in Class A Tolerances chart
  • Colorless graduation is clearly legible against contrasting red background
  • With standard taper pennyhead stoppers
Compliance
Designed from requirements for Class A unserialized flasks of ASTM™ E 288
Specifications
Specifications
Calibrated To Contain
1000 mL
13.3 in.
±0.3 mL
22
Glass
Borosilicate Glass
33.8 cm
Yes