Kimble™ KIMAX™ Flasks with Detachable Side Arm
Detachable plastic side arm secured by neoprene grommet
Manufacturer:  Kimble™ K270654000
 View more versions of this product
Catalog No. 10-181-5E
Specifications Description & Specifications
Specifications
Yes
12
20.8cm
250mL
38cm
4000mL
ASTM E 1406, Type III, Class 2
8.18 in.
1000 to 4000mL
14.96 in.
Detachable side arm
Borosilicate Glass
Accessories