Kimble™ KIMAX™ Distilling Column
$227.70 - $246.20
Includes: Supplied with jacket to make column a condenser.
Products ${productFamilyLength}
Catalog Number Mfr. No. Price Quantity  
Catalog Number Mfr. No. Price Quantity  
10-310-55 Kimble™
K21805300
Case for $246.20
11-301-49 Kimble™
K21802200
Case for $227.70
Specifications
Description
  • Outer standard taperjoint at top; inner standard taperjoint with drip tip at bottom
  • Two connections for 5/16 in. I.D. (8mm) flexible tubing
  • Indentation ring near bottom of inner tube supports packing material