Kimble™ KIMAX™ Class B Flasks with standard taper Pennyhead Stopper
Calibrated “To Contain” within Class B tolerances
Manufacturer:  Kimble™ K2801510
 View more versions of this product
Catalog No. 10-204A
Specifications Description & Specifications
Specifications
Calibrated To Contain
9
10.3cm
10mL
Glass
4.05 in.
±0.04mL
Borosilicate Glass