Please login to your online account to display your discounted pricing

Kimble™ KIMAX™ Caps for Media Bottles


Black phenolic or propylene caps

Manufacturer: kimble™  K7380838430

 View more versions of this product

Catalog No. 03-340-31

  • / Case of 48 Each
 

Description & Specifications

Specifications

For Use With (Equipment) Media bottle
Neck Finish 38-430
Color Black
Material Polypropylene
No. per Case 48 Cs.
Closure Liner PTFE/Silicone