DWK Life Sciences Kimble™ KIMAX™ Caps for Media Bottles
Black phenolic or propylene caps
Manufacturer:  DWK Life Sciences 7380338430
 View more versions of this product
Catalog No. 06-422-2B
Description
Description
  • Phenolic caps with plain 14B rubber liners or PTFE-faced 14B rubber liners
  • Polypropylene caps with bonded PTFE-faced silicone liners to eliminate glue contamination and loose liners
Specifications
Specifications
14B Rubber
Black
144 Cs.
Media bottle
Phenolic
38-430