DWK Life Sciences Kimble™ KIMAX™ Accuform Micro-Vial Graduated Aluminum Seal Vials
ACCUFORM™ micro-vials have a V-shaped interior to enable recovery of a minute residual sample by means of a syringe needle. Closures not included. Available in 2mL, 3mL and 5mL capacity.
Manufacturer:  DWK Life Sciences 607202
 View more versions of this product
Catalog No. 06-100-2D
Description
Description
ACCUFORM™ micro-vials have a V-shaped interior to enable recovery of a minute residual sample by means of a syringe needle. Closures not included. Available in 2mL, 3mL and 5mL capacity.
  • Graduated ACCUFORM™ Aluminum Seal Micro-Vials without Closures
  • Designed for an aluminum seal
  • Options:
    • 2mL Capacity—Height 40mm, OD 21mm
    • 3mL Capacity—Height 51mm, OD 21mm
    • 5mL Capacity—Height 62mm, OD 21mm
  • A/S Finish 20mm
  • Case of 12
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Specifications
Specifications
Clear
Borosilicate Glass
Enable recovery of a minute residual sample by means of a syringe needle
1.57 in.
Screw Cap
12 Case
21 mm
2 mL
Yes
Aluminum
40 mm