DWK Life Sciences Kimble™ KIMAX™ 5-Place NMR Tube Washers
These complete five-place washers use flexible PTFE tubing and a vacuum source (not included) to direct a power stream of wash solvent to thoroughly clean the insides of 7 and 8 in. NMR tubes.
Manufacturer:  DWK Life Sciences 8970330003
 View more versions of this product
Catalog No. K8970330003
Description
Description
These complete five-place washers use flexible PTFE tubing and a vacuum source (not included) to direct a power stream of wash solvent to thoroughly clean the insides of 7 and 8 in. NMR tubes.
  • Two sizes available for 3 and 5mm diameter tubes
  • Both sizes come with a 1L reservoir bottle
  • All wetted parts are constructed of chemical-resistant PTFE or borosilicate glass
  • An O-ring slips over each tube to form a vacuum-tight seal before tubes are positioned for cleaning
  • With a simple twist, the wash valve can be configured to wash and then dry
  • Cycles can be repeated, or cleaned tubes can be removed immediately for use
Specifications
Specifications
3mm dia. Tubes
PTFE or Borosilicate Glass
NMR Tube Washer