DWK Life Sciences Kimble™ ClearScrew Thread Sample Vials without Closures
Packaged in corrugated trays with partitions
Manufacturer:  DWK Life Sciences K608122857
 View more versions of this product
Catalog No. 14-957-229
Description
Description
  • Manufactured from clear 33 expansion borosilicate glass
Specifications
Specifications
ASTM Type I, Class A
Screw
24-400
Sample Vial
Clear
Borosilicate Glass
24-400
57mm
Screw
5.4 dr.
28 mm
20 mL
Documents