missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
DWK Life Sciences Kimble™ ClearScrew Thread Sample Vials with Attached PTFE-Faced/White Rubber Lined Caps
Packaged in corrugated trays with partitions
Description
- Small case quantities
- Manufactured from clear 33 expansion, borosilicate glass
- Attached closed top black phenolic closures
Specifications
A | |
Clear | |
Borosilicate Glass | |
Screw | |
Phenolic | |
85 mm |
PTFE/Rubber | |
23 mm | |
24 mL | |
6.5 dr. | |
Screw Cap (Closed-top) |