DWK Life Sciences Kimble™ BEAKERplus™ Combination Beakers and Flasks
Combination design to reduce splashing during agitation with funnels or filters
Supplier: DWK Life Sciences 14035150
Description
- Made from KG-33 borosilicate glass
- Graduated
- Wide fluted rim
- Pour spout
- Narrow recessed neck
- Chemical and heat resistant
Specifications
2.75 in. | |
Borosilicate Glass | |
8.3 cm | |
70 mm |
150 mL | |
3.26 in. | |
Yes |