DWK Life Sciences Kimble™ AmberScrew Thread Vials without Closures
Manufactured from amber 33 expansion borosilicate glass
Manufacturer:  DWK Life Sciences K609121940
 View more versions of this product
Catalog No. 14-957-267
Description
Description
  • Amber vials offer protection against harmful light and UV rays for light sensitive products.
Specifications
Specifications
USP Type I
Screw
15-425
Sample Vial
Amber
Borosilicate Glass
15-425
40mm
Screw
1.5 dr.
19 mm
5.5 mL
Documents