DWK Life Sciences Kimble™ AmberScrew Thread Vials with Attached White Rubber Lined Closures
Attached closed top black phenolic closures
Manufacturer:  DWK Life Sciences K60911D5
 View more versions of this product
Catalog No. 14-957-258
Description
Description
  • Manufactured from amber 51 expansion borosilicate glass
  • Closures with white rubber liners
  • Vials packaged in corrugated trays with partitions
Specifications
Specifications
USP Type I
Closed screw top
Rubber
Sample Vial
Amber
Borosilicate Glass
Phenolic
57mm
Screw
5.4 dr.
28 mm
20 mL
Documents