DWK Life Sciences Kimble™ AmberScrew Thread Vials with Attached White Rubber Lined Closures
Attached closed top black phenolic closures
Manufacturer:  DWK Life Sciences K60911D12
 View more versions of this product
Catalog No. 14-957-256
Description
Description
  • Manufactured from amber 51 expansion borosilicate glass
  • Closures with white rubber liners
  • Vials packaged in corrugated trays with partitions
Specifications
Specifications
USP Type I
Screw cap
Rubber
Sample Vial
Amber
Borosilicate Glass
Phenolic
35mm
Screw
0.37 dr.
12mm
1.8mL
Documents