Please login to your online account to display your discounted pricing

Kimble™ AmberScrew Thread Vials with Attached Caps, 0.06in. Silicone Septa

Closures supplied with PTFE-faced, silicone septa

$337.12 - $337.12

Specifications

Closure Type Open screw top
Diameter (Metric) Outside 28mm
Color Amber
Material Borosilicate Glass
Type Sample Vial
View More Specs
Products
Catalog Number Mfr. No. Height (Metric) Capacity (English) Volume (Metric) Price Quantity    

14-957-260

 
kimble™
K60911S5
57mm 5 dr. 20mL Case of 72 Each for $337.12
Description & Specifications

Specifications

Closure Type Open screw top
Diameter (Metric) Outside 28mm
Color Amber
Material Borosilicate Glass
Type Sample Vial
Material Closure Polypropylene
Material (Septa) PTFE/Silicone
Thread Style Screw

  • Manufactured from amber 33 expansion borosilicate glass
  • Open top white polypropylene closures attached
  • Vials packaged in corrugated trays with partitions
  • PTFE Thickness: 0.005 in.
  • Silicone Rubber Thickness: 0.06 in.