DWK Life Sciences Kimble™ Aluminum Seal Caps
With PTFE-faced rubber septa
$355.56 - $554.08
Specifications
For Use With (Application) | Multiple injection |
---|---|
Material | Aluminum |
Neck Finish | 11 |
Catalog Number | Mfr. No. | Neck Finish | Color | No. per Case | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Neck Finish | Color | No. per Case | Price | Quantity | |||
03-341-601
|
DWK Life Sciences
N7382411 |
11 | Natural | 100 Cs. |
Case of 100 Each for $355.56
|
|
|||
03-341-602
|
DWK Life Sciences
N7382611 |
11 | Red | 1000 Cs. |
Case of 1000 Each for $554.08
|
|
|||
Description
- Moderate to good resealability
- One piece
- PTFE-faced red rubber septa
- For general GC applications
- Moderate resealability
- Not recommended for multiple injections
- PTFE-faced natural rubber septa
- Good resealability
- Multiple injections
Specifications
Multiple injection | |
Aluminum | |
11 |