Please login to your online account to display your discounted pricing

Kimble™ Aluminum Seal Caps


With PTFE-faced rubber septa

Manufacturer: kimble™  KN7382613

 View more versions of this product

Catalog No. 03-341-603

  • / Case of 1000 Each
 

Description & Specifications

Specifications

Closure Liner PTFE/Rubber
Color Red
For Use With (Application) Multiple injection
No. per Case 1000 Cs.
Neck Finish 13