Please login to your online account to display your discounted pricing

Kimble™ Brand GL 45 Media/Storage Bottles Replacement Orange Polypropylene Cap and Clear Pour Ring


Manufacturer: kimble™  K14395P45

 View more versions of this product

Catalog No. 02-542-3

  • / Case of 10 Each
 

Description & Specifications

Specifications

Type Pour ring
Neck Finish GL45
For Use With (Equipment) Media bottle
Color Clear
No. per Case 10 Cs.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.