Please login to your online account to display your discounted pricing

Kimble™ Black Phenolic Unlined Screw Thread Closures with Open Tops


Open top closures providing excellent chemical resistance

Manufacturer: kimble™  K7380424400

 View more versions of this product

Catalog No. 03-340-7K

  • / Case of 144 Each
 

Description & Specifications

Specifications

Color Black
For Use With Bottle
Material Phenolic
Neck Finish 24-400
No. per Case 144 Cs.