missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

DWK Life Sciences Kimble™ Black Phenolic Unlined Screw Thread Closures with Open Tops

Open top closures providing excellent chemical resistance

$80.29 - $308.91

Specifications

Color Black
Material Phenolic
For Use With (Equipment) Bottle
No. per Case 144 Cs.
View More Specs

Products 4
Catalog Number Mfr. No. Neck Finish Price Quantity  
Catalog Number Mfr. No. Neck Finish Price Quantity  
03-340-7E
View Documents Promotion Details
DWK Life Sciences
7380415425
15-425
Case of 144 Each for $80.29
 
03-340-7G
View Documents Promotion Details
DWK Life Sciences
7380418400
18-400
Case of 144 Each for $85.59
 
03-340-7K
View Documents Promotion Details
DWK Life Sciences
7380424400
24-400
Case of 144 Each for $100.41
 
03-340-7J
View Documents Promotion Details
DWK Life Sciences
7380420400
20-400
Case of 144 Each for $308.91
 
Description

Description

Open top closures providing excellent chemical resistance

  • Phenolic
  • Open top
  • Autoclavable
  • Unlined
  • Black

Specifications

Specifications

Black
Phenolic
Bottle
144 Cs.
SDS
Product Certifications