Please login to your online account to display your discounted pricing

Kimble™ Black with Cone-Shaped Liner Phenolic Caps/Closures


Black caps and closures offer excellent sample storage and re-sealing properties

Manufacturer: kimble™  7380918400

 View more versions of this product

Catalog No. 03-340-208

  • / Case of 144 Each
 

Description & Specifications

Specifications

Thread Size 18-400
Type Solid Top Screw Cap
Material (Liner) Polyethylene
Color Black
GPI Thread 18-400
Material Closure Phenolic
For Use With (Application) Excellent sample storage and re-sealing properties
Material Phenolic