Please login to your online account to display your discounted pricing
Kimble™ Amber Glass Wide-Mouth Packers with PTFE-Faced LDPE Foam-lined Caps
Amber protects light-sensitive contents
$65.82 - $126.36
Specifications
Closure Liner PTFE/LDPE
Closure Material Phenolic
Closure Screw
Closure Color Black
Color Amber
View More Specs
Products
Catalog Number Mfr. No. Capacity Diameter (English) Height (English) Closure Size Price Quantity    
13-756-501
kimble™
K5220845V26
8 oz., 250mL 2.59 in. 4.68 in. 45-400 Case of 12 Each for $65.82
13-756-499
kimble™
K5220233V26
2 oz., 60mL 1.73 in. 2.95 in. 33-400 Case of 24 Each for $109.85
13-756-503
kimble™
K5223253V26
32 oz., 1000mL 3.89 in. 7 in. 53-400 Case of 12 Each for $126.36
13-756-502
kimble™
K5221653V26
16 oz., 500mL 3.14 in. 5.75 in. 53-400 Case of 12 Each for $94.69
Description & Specifications
Specifications
Closure Liner PTFE/LDPE
Closure Material Phenolic
Closure Screw
Closure Color Black
Color Amber
Material Glass
All 32-ounce jars are packed in corrugated cartons with divider cells.