Kimble™ Amber Glass Jugs without Caps
$84.97 - $96.39
Specifications
Closure Size 38-400
Color Amber
Material Soda Lime Glass
Mouth Narrow
Includes Sturdy handle,Sturdy handle
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Diameter (English) Height (English) Capacity (English) Price Quantity  
Catalog Number Mfr. No. Diameter (English) Height (English) Capacity (English) Price Quantity  
13-757-209 Kimble™
K5929138B
6.22 in. 13.4 in. 130 oz. Case of 4 Each for $84.97
13-757-208 Dwk Life Sciences Llc
K5928038B
5.39 in. 12.2 in. 80 oz. Case of 6 Each for $96.39
Specifications
Description
  • Narrow mouth and sturdy handle for safe pouring
  • Amber protects light-sensitive contents
  • Type III soda-lime glass
All jugs are packed in corrugated cartons with divider cells.
Description & Specifications
Specifications
38-400
Soda Lime Glass
Sturdy handle,Sturdy handle
Amber
Narrow