Kimble™ Amber Glass Jugs: Caps with Taperseal
$82.63 - $125.56
Specifications
Closure Liner Taperseal
Closure Size 38-400
Color Amber
Material Soda Lime Glass
Mouth Narrow
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Diameter (English) Height (English) Capacity (English) Price Quantity  
Catalog Number Mfr. No. Diameter (English) Height (English) Capacity (English) Price Quantity  
13-757-202 Kimble™
K5928038V25
5.39 in. 12.2 in. 80 oz. Case of 6 Each for $125.56
13-757-203 Kimble™
K5929138V25
6.22 in. 13.4 in. 130 oz. Case of 4 Each for $82.63
Specifications
Description
  • Narrow mouth and sturdy handle for safe pouring
  • Amber protects light-sensitive contents
  • Black phenolic caps
  • Type III soda-lime glass
All jugs are packed in corrugated cartons with divider cells (caps attached).
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Description & Specifications
Specifications
Taperseal
Amber
Narrow
Sturdy handle
38-400
Soda Lime Glass
Cylindrical