Kimble™ Amber Glass Jugs: Caps with PTFE-Faced LDPE Foam
$85.82 - $121.38
Specifications
Closure Liner PTFE/LDPE
Closure Size 38-400
Color Amber
Material Soda Lime Glass
Mouth Narrow
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Diameter (English) Height (English) Capacity (English) Price Quantity  
Catalog Number Mfr. No. Diameter (English) Height (English) Capacity (English) Price Quantity  
13-757-206 Kimble™
K5928038V26
5.39 in. 12.2 in. 80 oz. Case of 6 Each for $121.38
13-757-207 Kimble™
K5929138V26
6.22 in. 13.4 in. 130 oz. Case of 4 Each for $85.82
Specifications
Description
  • Narrow mouth and sturdy handle for safe pouring
  • Amber protects light-sensitive contents
  • Black phenolic caps
  • Type III soda-lime glass
All jugs are packed in corrugated cartons with divider cells (caps attached).
Description & Specifications
Specifications
PTFE/LDPE
Amber
Narrow
Sturdy handle
38-400
Soda Lime Glass
Cylindrical