Please login to your online account to display your discounted pricing
Kimble™ Aluminum Seals for Headspace Vials
Allow for easy access to vial contents
Manufacturer: kimble™  K7382120
 View more versions of this product
Catalog No. 03-340-28
  • / Case of 1000 Each
Description & Specifications
Specifications
Quantity 1000 Case
Type One Piece Seal
Color Natural
Material Closure Aluminum
Thread Size 20mm
Closure Material Aluminum
Closure Type Tear-off
Accessories
Accessories for Catalog Number: 0334028
Item Mfr. No. Item Description Price Per Unit Quantity  
03-375-24A
Thermo Scientific™ Manual Crimpers, Decrimpers and Decrimping Pliers
thermo scientific™
C4020100
Achieve secure, repeatable crimps or remove seals with minimized risk of damage. Each for $469.08