missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Kimble™ 20mL Glass Screw-Thread Scintillation Vials

Manufactured from clear KG-33 borosilicate glass

Manufacturer:  Kimble™ K7451520

 View more versions of this product

Catalog No. 03-340-4F


 

Specifications Description & Specifications

Specifications

0.67 oz., 20mL
22mm Unattached Screw cap
White Urea
Cellular trays
20mL
Borosilicate Glass
USP Type I, ASTM E438
Fast and easy identification
Poly-Seal cone
0.67 oz.
White